Pharmaceutical secondary standard, glycolic acid, CRM, Supelco®
Catalog # PHR1427-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:Pharmaceutical standards
- CAS No.:79-14-1
- Description:Glycolic acid (hydroxyacetic acid)
- Formula:C₂H₄O₃
- Description:Glycolic acid (hydroxyacetic acid)
Specifications
About this item
This secondary standard offers multi-traceability to the USP, EP and BP primary standards, where they are available.
Certifications: In accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDES 34:2009 (AClass Cert AR-1470).