

Acrylamide, Supelco®
Catalog # 23701-100MG-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:71.08 g/mol
Boiling Pt:232 °C (1013 hPa)
Melting Pt:84.5 °C
Density:1.122 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:2074
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS06
:GHS08
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032