



2-Methylbutane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-78-4
:
Formula:CH₃CH₂CH(CH₃)₂
MDL Number:MFCD00009338
:72.15 g/mol
Boiling Pt:28 °C (1013 hPa)
Melting Pt:–160 °C
Density:0.62 g/cm³ (20 °C)
Flash Pt:–51 °C
UN:1265
ADR:3,I
:M-Clarity™ quality level = MQ100
EINECS:201-142-8
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together