Supelco® Organic Reference Standard, 3-Methylbutanol
59092-5ML
: Merck
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:123-51-3
- Description:3-Methylbutanol
- Seq:02
- Storage conditions:2...30 °C
- Hazard class description:H226 - H315 - H318 - H332 - H335
- Shipping conditions:STD
- MDL No.:MFCD00002934
- M-Clarity quality level:MQ100
- Synonym(s):3-Methyl-1-butanol; Isoamyl alcohol; Isopentyl alcohol
- Formula:(CH₃)₂CHCH₂CH₂OH
- Molecular weight:88,15
- Density:0,809 g/ml at 20 °C (lit.)
- Flash point:43,5 °C - Closed cup
- Boiling point:131 - 132 °C
- GHS signal: GHS02, GHS05, GHS07
: P210 - P233 - P280 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338