


Isobutanol ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:78-83-1
:
:(CH₃)₂CHCH₂OH
:MFCD00004740
:74.12 g/mol
:13,05148
:107.9 °C (1013 hPa)
:–108 °C
:0.802 g/cm³ (20 °C)
:28 °C
:Ambient
:201-148-0
:1212
:3,III
:GHS02
:GHS05
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Assay:Min. 99.0 %
- Acidity:Max. 0.0006 meq/g
- Boiling point:107 - 109 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.799 - 0.803
- Density (20/20):0.800 - 0.804
- Refractive index (15°C):1.397 - 1.399
- Refractive index (20°C):1.395 - 1.396
- Evaporation residue:Max. 50 ppm
- Water:Max. 0.5 %
- Conforms to Reag. Ph.Eur.:Passes test
Frequently Bought Together