AMP (2-Amino-2-methylpropanol), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP
CAS Number:124-68-5
:
Formula:(CH₃)₂C(NH₂)CH₂OH
MDL Number:MFCD00008051
Boiling Pt:165
Melting Pt:18…26 °C
Density:0.667 g/cm³ (20 °C)
Molecular Weight:89.14 g/mol
Flash Pt:67
Merck Index:13,00448
:M-Clarity™ quality level = MQ200
EINECS:204-709-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together