
2-Ethylcaproic acid, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:149-57-5
:
Formula:C₈H₁₆O₂
MDL Number:MFCD00002675
Boiling Pt:228 °C (1013 hPa)
Melting Pt:–82.8 °C
Density:0.91 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:114 °C
:144.21 g/mol
EINECS:205-743-6
:GHS08
Storage Temperature:Ambient
Frequently Bought Together