Ethanol 96% ≥96% (v/v), TechniSolv®, pure
83804.930
:
- Pk:950 L
- Seq:5783
- Environmentally Preferable:
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
: Ethyl alcohol
:
:46.07 g/mol
:12 °C (closed cup)
:200-578-6
:1170
:01-2119457610-43
:GHS02
:GHS07
:Ambient
:64-17-5
:C₂H₅OH
:MFCD00003568
:78.3 °C (1013 hPa)
:–117 °C
:0.7895 g/cm³ (20 °C)
:3 II
:12,03806