


1-Butanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
:M-Clarity™ quality level = MQ300
Flash Pt:30 °C
REACH:01-2119484630-38
:GHS02
:GHS05
:GHS07
Frequently Bought Together