
Propionic anhydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-62-6
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00009303
:130.14 g/mol
Merck Index:12,08011
Boiling Pt:167 °C (1013 hPa)
Melting Pt:–45 °C
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-638-2
UN:2496
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:64 °C
:GHS05
Frequently Bought Together