2-Ethoxyethanol ≥99.5%, AnalaR NORMAPUR® analytical reagent
103426R
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Assay:Min. 99.5 %
- IR Spectrum:Passes test
- Acidity:Max. 0.0030 meq/g
- Colouration:Max. 20 APHA
- Density (20/4):0.929 - 0.930
- Evaporation residue:Max. 100 ppm
- Peroxides (as H2O2):Max. 0.02 %
- Water:Max. 0.1 %
- Al (Aluminium):Max. 10 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 2 ppm
- Ni (Nickel):Max. 0.05 ppm
- Pb (Lead):Max. 0.5 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 50 ppm
:
:90.12 g/mol
:13,03786
:135 °C (1013 hPa)
:–70 °C
:0.932 g/cm³ (20 °C)
:40 °C (closed cup)
:Ambient
:203-804-1
:1171
:3,III
:01-2119560582-38
:GHS02
:GHS08
:GHS07
:110-80-5
:MFCD00008590
:C₂H₅OCH₂CH₂OH
:MFCD00002869