N,N,N',N'-Tetramethylethylenediamine (TEMED), Sigma-Aldrich®
: Merck
:110-18-9
:
:C₆H₁₆N₂
:MFCD00008335
:116.21 g/mol
:120 °C (1013 hPa)
:–64 °C
:0.7765 g/cm³ (20 °C)
:17 °C (closed cup)
:Ambient
:2372
:3,II
:M-Clarity™ quality level = MQ200
:203-744-6
:GHS02
:GHS05