Thioacetic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:507-09-5
:
Formula:C₂H₄OS
MDL Number:MFCD00004853
Boiling Pt:97 °C
Melting Pt:–17 °C
Density:1.068
Molecular Weight:76.12 g/mol
Flash Pt:18 °C (64 °F)
UN:2436
ADR:3 II
Merck Index:12,09454
EINECS:208-063-8
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together