Thioacetic acid 98%
:507-09-5
:
:C₂H₄OS
:MFCD00004853
:87…93 °C (760 mmHg)
:–17 °C
:1.07 g/cm³
:76.12 g/mol
:18 °C
:Refrigerator
:2436
:3 II
:12,09454
:208-063-8
:GHS06
:GHS05
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4610 to 1.4650 (20°C, 589 nm)
- Titration Argentometric:≥97.5 %
Frequently Bought Together