Citric acid, BAKER, J.T.Baker®
Catalog # 1995.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.5 - 101.0 %
- Appearance of solution:passes test
- Heavy Metals (as Pb):max. 10 ppm
- Identification:passes test
- Oxalic acid:max. 350 ppm
- Readily Carbonisable Substances:passes test
- Sulfated Ash:max. 0.1 %
- Sulfates (as SO4):max. 150 ppm
- Water (H2O):max. 1.0 %
Specification Test Results
About this item
:
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.665 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669