Potassium acetate, Sigma-Aldrich®
Catalog # SIALP1222-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Density:1.57 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>250 °C
Storage Temperature:Ambient
EINECS:204-822-2
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458