


Formic acid ≥96% ACS, Fluka™
Supplier: HONEYWELL
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Boiling Pt:100.4 °C (1.013 hPa)
Melting Pt:4 °C
Density:1.22 g/cm3 (20 °C)
Flash Pt:49.5 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8,II
:GHS05
:GHS06
:GHS02
Specification Test Results
- Vapor Pressure:44.8 mmHg ( 20 °C)
- Assay (alkalimetric):Min. 96 %
- Identity (IR):complying
- Non-volatile matter:Max. 0.003 %
- Ammonium (NH₄):Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.001 %
- Sulfate (SO₄):Max. 0.003 %
- Sulfite (SO₃):complying
- Acetic acid (CH₃COOH):Max. 0.4 %
- APHA:Max. 15
- Appearance:complying
- Solution 1 mL + 3 mL (H₂O):complying
Specification Test Results
Frequently Bought Together