Diethyl malonate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105-53-3
:
Formula:C₇H₁₂O₄
MDL Number:MFCD00009195
:160.17 g/mol
Merck Index:13,03856
Boiling Pt:199 °C (760 mmHg)
Melting Pt:–50 °C
Density:1.055 g/cm³
Flash Pt:93 °C
Storage Temperature:Ambient
EINECS:203-305-9
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4160 (20°C, 589 nm)
- Density:(20°C) 1.052 to 1.058 g/ml
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together