

Ethyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:141-78-6
:
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171
:88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
EINECS:205-500-4
UN:1173
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:–4 °C
REACH:01-2119475103-46
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together