Propiononitrile, Supelco®
Catalog # 76671-1ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:55.08 g/mol
Merck Index:12,08012
Boiling Pt:97.2 °C (1013 hPa)
Melting Pt:–93 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:203-464-4
UN:2404
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:107-12-0
Formula:C₃H₅N
MDL Number:MFCD00001948