Chloroacetamide
Glass bottle for solids, 100 mg
Catalog # PROOMM0083.06
Supplier: PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 2-Chloroacetamide
- CAS Number:79-07-2
- Formula:C₂H₄ClNO
- MDL Number:MFCD00008027
- Molecular Weight:93.51 g/mol
- Merck Index:13,02127
- Boiling Pt:256 °C (1013 hPa)
- Melting Pt:115…117 °C
- Density:1.6 g/cm³ (20 °C)
- Flash Pt:170 °C
- Storage Temperature:Ambient
- EINECS:201-174-2
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



