3-Bromo-1-propanol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-18-9
:
Formula:BrH₂CCH₂CH₂OH
MDL Number:MFCD00002942
:138.99 g/mol
Boiling Pt:71…73 °C (11 mmHg)
Density:1.590
Flash Pt:93 °C (199 °F)
EINECS:210-986-6
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together