

1,10-Phenanthroline monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5144-89-8
:
Formula:C₁₂H₈N₂·1H₂O
MDL Number:MFCD00149973
:198.22 g/mol
Merck Index:13,07291
Melting Pt:93…94 °C
Density:1.259 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
:GHS06
:GHS09
Frequently Bought Together