1,10-Phenanthroline monohydrate ≥99.0% analytical reagent
Catalog # 26227.101
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Suited for iron reagent:Passes test
- Melting point:93 - 110 °C
- Ignition residue (SO4):Max. 0.1 %
Specification Test Results
About this item
:
:198.22 g/mol
Merck Index:13,07291
Melting Pt:93…94 °C
Density:1.259 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-629-2
UN:2811
ADR:6.1,III
:GHS06
:GHS09
CAS Number:5144-89-8
Formula:C₁₂H₈N₂·1H₂O
MDL Number:MFCD00149973