D-(-)-Sorbitol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
:182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
Frequently Bought Together