L(+)-Tartaric acid, BioXtra, Sigma-Aldrich®
: Merck
:87-69-4
:
:C₄H₆O₆
:MFCD00064207
:13,09158
:399 °C (1013 hPa)
:168…170 °C
:1.759 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:150 °C (closed cup)
:150.09 g/mol
:Ambient
:201-766-0
:01-2119537204-47
:GHS05
Frequently Bought Together