Order Entry
Slovakia
ContactUsLinkComponent
Pesticide reference standard, (±)-α-BHC 1,000 µg/ml in methanol, Supelco®
Pesticide reference standard, (±)-α-BHC 1,000 µg/ml in methanol, Supelco®
  40100-U
 :  Merck
 :  
Pesticide reference standard, (±)-α-BHC 1,000 µg/ml in methanol, Supelco®
  40100-U
 :  Merck
 :  

 

  • Pk:
    1 mL
  • CAS No.:
    319-84-6
  • Description:
    (±)-α-BHC 1,000 µg/ml in methanol
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₆H₆Cl₆
  • Description:
    (±)-α-BHC