Organic reference standard, 2,3,3',4-Tetrachlorobiphenyl (PCB No. 55) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20005500IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:74338-24-2
- Description:2,3,3',4-Tetrachlorobiphenyl (PCB No. 55) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3,3',4-Tetrachlorobiphenyl (PCB No. 55)