Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, 2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-121
Supplier:  PROMOCHEM
CAS Number:  
Organic reference standard, 2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-121
Supplier:  PROMOCHEM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    56558-18-0
  • Description:
    2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3',4,5',6-Pentachlorobiphenyl (PCB No. 121)

Specifications

About this item