Sodium gluconate, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:527-07-1
:
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210
Merck Index:12,08766
Melting Pt:170…175 °C
:218.14 g/mol
Storage Temperature:Ambient
EINECS:208-407-7
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
spec_VWRC27871.295 spec_VWRC27871.460
Product specification
Frequently Bought Together