Organic reference standard, 2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-123
Supplier: PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:65510-44-3
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123)