Order Entry
Slovakia
ContactUsLinkComponent
Manganese(II) sulphate monohydrate, Sigma-Aldrich®
Health
Environment
Manganese(II) sulphate monohydrate, Sigma-Aldrich®
Catalog # SIALM7899-500G
Supplier:  Merck
CAS Number:  
Health
Environment
Manganese(II) sulphate monohydrate, Sigma-Aldrich®
Catalog # SIALM7899-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Merck Index:13,05763
Melting Pt:700 °C
Density:2.95 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :169.02 g/mol
EINECS:232-089-9
 :GHS08
 :GHS09
CAS Number:10034-96-5
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159