Magnesium sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
Specification Test Results
- Assay:Min. 99.5 %
- Solution in water:Passes test
- Acidity or alkalinity:Max. 0.001 meq/g
- pH (25°C; 5 %):5.0 - 8.0
- Alkaline metals (as SO4):Max. 0.05 %
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Total N (Nitrogen):Max. 20 ppm
- Cl (Chloride):Max. 5 ppm
- NH4 (Ammonium):Max. 20 ppm
- NO3 (Nitrate):Max. 10 ppm
- PO4 (Phosphate):Max. 10 ppm
- As (Arsenic):Max. 2 ppm
- Ca (Calcium):Max. 50 ppm
- Cu (Copper):Max. 2 ppm
- Fe (Iron):Max. 1 ppm
- K (Potassium):Max. 10 ppm
- Mn (Manganese):Max. 5 ppm
- Na (Sodium):Max. 10 ppm
- Pb (Lead):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC25165.260 spec_VWRC25165.292 spec_VWRC25165.361 spec_VWRC25165.460 spec_VWRC25165.460
Product specification
More
Frequently Bought Together