Magnesium sulphate heptahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:14,5691
Melting Pt:1124 °C
Density:1.68
Storage Temperature:Ambient
EINECS:231-298-2
Related Information
Frequently Bought Together