Magnesium nitrate hexahydrate, Sigma-Aldrich®
: Merck
:13446-18-9
:10213-15-7
:Mg(NO₃)₂·6H₂O
:MFCD00149779
:256.41 g/mol
:13,05697
:330 °C (1013 hPa)
:89 °C
:1.63 g/cm³ (20 °C)
:Ambient
:233-826-7
:1474
:5.1,III
:M-Clarity™ quality level = MQ100
:93.3 °C
:GHS03
Frequently Bought Together