Sodium chromate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10034-82-9
:
Formula:CrNa₂O₄·4H₂O
MDL Number:MFCD00149167
:234.03 g/mol
Melting Pt:792.2 °C
Density:2.73 g/cm³ (20 °C)
EINECS:231-889-5
UN:3288
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
:GHS09
Frequently Bought Together