1-Octanesulphonic acid sodium salt monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207596-29-0
:
Formula:C₈H₁₇NaO₃S·1H₂O
MDL Number:MFCD00007544
:234.29 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:226-195-4
Related Information
Frequently Bought Together