Citric acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together