

Cadmium acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 22189.294
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Cl (Chloride):Max. 20 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ca (Calcium):Max. 50 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 5 ppm
- Na (Sodium):Max. 20 ppm
- Pb (Lead):Max. 50 ppm
- Zn (Zinc):Max. 20 ppm
Specification Test Results
About this item
:
:266.53 g/mol
Merck Index:13,01614
Boiling Pt:105…107 °C (4 Torr)
Melting Pt:256 °C
Density:2.341 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-853-2
UN:2570
ADR:6.1,III
:GHS07
:GHS09
CAS Number:5743-04-4
Formula:(H₃CCOO)₂Cd·2H₂O
MDL Number:MFCD00150020