Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Guanidinium hydrochloride
Guanidinium hydrochloride, Sigma-Aldrich®
Guanidinium hydrochloride, Sigma-Aldrich®
:
Merck
Guanidinium hydrochloride, Sigma-Aldrich®
:
Merck
:
50-01-1
:
:
NH₂C(=NH)NH₂·HCl
:
MFCD00013026
:
95.53 g/mol
:
13,04578
:
185 °C
:
1.354 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ400
:
200-002-3
:
GHS07
:
Ambient
SIALG4505-500G_TDS_EN
Frequently Bought Together
(±)-1-Methoxy-2-propanol, EMPLURA®, Supelco®
: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Ethyl acetate ≥99%, TECHNICAL
: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Butyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
(±)-2-Methyltetrahydrofuran, EMPLURA®
: Merck
Triethylamine, Sigma-Aldrich®
: Merck