Spermidine trihydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride
CAS Number:334-50-9
:
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918
:254.63 g/mol
Merck Index:12,08893
Melting Pt:256…258 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
EINECS:206-379-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together