Order Entry
Slovakia
ContactUsLinkComponent
Glutaraldehyde 50% (w/w) in water
Glutaraldehyde 50% (w/w) in water
Certificates
undefined
Glutaraldehyde 50% (w/w) in water

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤50 APHA
  • Strength:
    50 to 51 %
  • pH:
    3.2 to 4.2
  • Methanol:
    ≤0.5 % (GC)

Specification Test Results

Related Information

Frequently Bought Together