Order Entry
Slovakia
ContactUsLinkComponent
DL-Leucine, Sigma-Aldrich®
DL-Leucine, Sigma-Aldrich®
Catalog # SIALL7875-25G
Supplier:  Merck
CAS Number:  
DL-Leucine, Sigma-Aldrich®
Catalog # SIALL7875-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :131.17 g/mol
Merck Index:13,05470
Boiling Pt:226 °C (1013 hPa)
Density:1.191 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-328-2
CAS Number:328-39-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD00063087