Order Entry
Slovakia
ContactUsLinkComponent
DL-Leucine
  EHERC14629300
 :  
DL-Leucine
  EHERC14629300
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
 :
 :131.17 g/mol
 :13,05470
 :226 °C (1013 hPa)
 :1.191 g/cm³ (20 °C)
 :Ambient
 :206-328-2
 :328-39-2
 :C₆H₁₃NO₂
 :MFCD00063087