DL-Methionine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:59-51-8
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063096
:149.21 g/mol
Merck Index:13,06004
Melting Pt:∼136 °C
Density:1.34 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:200-432-1
Frequently Bought Together