Order Entry
Slovakia
ContactUsLinkComponent
 
DL-Methionine, Sigma-Aldrich®
Catalog # SIALM2768-100G
Supplier:  Merck
CAS Number:  
DL-Methionine, Sigma-Aldrich®
Catalog # SIALM2768-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :149.21 g/mol
Merck Index:13,06004
Melting Pt:∼136 °C
Density:1.34 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-432-1
CAS Number:59-51-8
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063096