Bovine serum albumin (BSA), Fraction V
ROTH8076.4
:
- Pack type:Glass bottle for solids
- Pk:200 g
- CAS No.:90604-29-8
9048-46-8 - Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Form:powder
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:>98%
- Grade:for molecular biology