Agarose, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-731-8
6 Options Available Below
Product Details & Documents
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:232-731-8
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



