
Gentamicin sulphate for tissue culture
Catalog # 0304-10G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Loss on Drying:≤18%
- Methanol:≤1%
- pH (4%, Water) at 25 °C:3.5 - 5.5
- Potency (Dry Basis):≥590 mcg/mg
- Residue after Ignition:<1%
- Specific Rotation:107 - 121°
Specification Test Results
About this item
:
Merck Index:13,04403
Melting Pt:218…237 °C
EINECS:215-778-9
:GHS08
Storage Temperature:Refrigerator
CAS Number:1405-41-0
MDL Number:MFCD00270181