

Tetrahydrofuran-[D₈], Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1693-74-9
:
Formula:C₄D₈O
MDL Number:MFCD00044238
:80.04 g/mol
Boiling Pt:65 °C (1013 hPa)
Melting Pt:–108 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:–17 °C (closed cup)
UN:2056
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:216-898-4
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together